missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LGALS13 Full-length ORF (AAH66304.1, 1 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00029124-P02.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129062

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene has lysophospholipase activity. It is composed of two identical subunits which are held together by disulfide bonds. This protein has structural similarity to several members of the beta-galactoside-binding S-type lectin family. [provided by RefSeq]

Sequence: MSSLPVPYKLPVSLSVGSCVIIKGTPIHSFINDPQLQVDFYTDMDEDSDIAFRFRVHFGNHVVMNRREFGIWMLEETTDYVPFEDGKQFELCIYVHYNEYEIKVNGIRIYGFVHRIPPSFVKMVQVSRDISLTSVCVCN
Specifica

Specifica

AAH66304.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
42.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GAL13/PLAC8/PP13
LGALS13
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
29124
LGALS13 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSLPVPYKLPVSLSVGSCVIIKGTPIHSFINDPQLQVDFYTDMDEDSDIAFRFRVHFGNHVVMNRREFGIWMLEETTDYVPFEDGKQFELCIYVHYNEYEIKVNGIRIYGFVHRIPPSFVKMVQVSRDISLTSVCVCN
RUO
LGALS13
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali