missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LOC440456 Full-length ORF (ENSP00000313104, 1 a.a. - 237 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00440456-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: MTSPRTGTPALDNQPQTGIPRAAGGRDAGTPGQQIPGGEGKGEGGTGEAHPPWAPLATSQRPFRRSLPGSGPRLSAPVLVPMPPKVPSRGIQVPPRPAQTQLLWVWVTALLSVAVPIRCRCQLRFRETSPAPPRKPRPGRWSGPNLPAPPTRPLGSLRPAQQTRGTLDKVAKEPYGSPGLDPAPSVPQPRQAAVSWPLAWVTLGGRRRCGQCGSLEGPCTSGEDHRLMALVEGRRQISpecifica
ENSP00000313104 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PLEKHM1P | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
440456 | |
LOC440456 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTSPRTGTPALDNQPQTGIPRAAGGRDAGTPGQQIPGGEGKGEGGTGEAHPPWAPLATSQRPFRRSLPGSGPRLSAPVLVPMPPKVPSRGIQVPPRPAQTQLLWVWVTALLSVAVPIRCRCQLRFRETSPAPPRKPRPGRWSGPNLPAPPTRPLGSLRPAQQTRGTLDKVAKEPYGSPGLDPAPSVPQPRQAAVSWPLAWVTLGGRRRCGQCGSLEGPCTSGEDHRLMALVEGRRQI | |
RUO | |
PLEKHM1P | |
Recombinant | |
wheat germ expression system |