missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LRP1B Partial ORF (NP_061027, 111 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00053353-Q01.25ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
LRP1B belongs to the low density lipoprotein (LDL) receptor gene family. These receptors play a wide variety of roles in normal cell function and development due to their interactions with multiple ligands (Liu et al., 2001 [PubMed 11384978]).[supplied by OMIM]
Sequence: VHCQELLSNCQQLNCQYKCTMVRNSTRCYCEDGFEITEDGRSCKDQDECAVYGTCSQTCRNTHGSYTCSCVEGYLMQPDNRSCKAKIEPTSpecifica
NP_061027 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VHCQELLSNCQQLNCQYKCTMVRNSTRCYCEDGFEITEDGRSCKDQDECAVYGTCSQTCRNTHGSYTCSCVEGYLMQPDNRSCKAKIEPT | |
RUO | |
LRP1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
53353 | |
LRP1B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LRP-DIT/LRPDIT | |
LRP1B | |
Recombinant | |
wheat germ expression system |