missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LUC7L Full-length ORF (NP_060502.1, 1 a.a. - 325 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00055692-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16131693

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The LUC7L gene may represent a mammalian heterochromatic gene, encoding a putative RNA-binding protein similar to the yeast Luc7p subunit of the U1 snRNP splicing complex that is normally required for 5-prime splice site selection (Tufarelli et al., 2001 [PubMed 11170747]).[supplied by OMIM]

Sequence: MSAQAQMRALLDQLMGTARDGDETRQRVKFTDDRVCKSHLLDCCPHDILAGTRMDLGECTKIHDLALRADYEIASKERDLFFELDAMDHLESFIAECDRRTELAKKRLAETQEEISAEVSAKAEKVHELNEEIGKLLAKAEQLGAEGNVDESQKILMEVEKVRAKKKEAEEEYRNSMPASSFQQQKLRVCEVCSAYLGLHDNDRRLADHFGGKLHLGFIQIREKLDQLRKTVAEKQEKRNQDRLRRREEREREERLSRRSGSRTRDRRRSRSRDRRRRRSRSTSRERRKLSRSRSRDRHRRHRSRSRSHSRGHRRASRDRSAKYK
Specifica

Specifica

NP_060502.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10231/LUC7-LIKE/LUC7B1/SR+89
LUC7L
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55692
LUC7L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSAQAQMRALLDQLMGTARDGDETRQRVKFTDDRVCKSHLLDCCPHDILAGTRMDLGECTKIHDLALRADYEIASKERDLFFELDAMDHLESFIAECDRRTELAKKRLAETQEEISAEVSAKAEKVHELNEEIGKLLAKAEQLGAEGNVDESQKILMEVEKVRAKKKEAEEEYRNSMPASSFQQQKLRVCEVCSAYLGLHDNDRRLADHFGGKLHLGFIQIREKLDQLRKTVAEKQEKRNQDRLRRREEREREERLSRRSGSRTRDRRRSRSRDRRRRRSRSTSRERRKLSRSRSRDRHRRHRSRSRSHSRGHRRASRDRSAKYK
RUO
LUC7L
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali