missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human M-CSF R/CD115 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB3292-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifica
| M-CSFR/CD115 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1436 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 61715 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P07333.2 | |
| CSF1R | |
| Mouse myeloma cell line NS0-derived recombinant human M-CSF R/CD115 Ile20-Glu512 (Pro54Ala) Accession # P07333.2 | |
| 25 μg | |
| Primary | |
| Detects human M-CSF R/CD115 in ELISAs. In ELISAs, no cross-react with recombinant human (rh) M-CSF, rhGM-CSF, rhPDGF R alpha , rhPDGF R beta , or rmM-CSF is observed. | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto