missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human M-CSF R/CD115 Antibody, R&D Systems™

Mouse Monoclonal Antibody
242.00€ - 2055.00€
Specifica
| Antigene | M-CSFR/CD115 |
|---|---|
| Clone | 61715 |
| Diluizione | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applicazioni | ELISA |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
18798778
![]() |
R&D Systems
MAB3292-SP |
25 μg |
242.00€
25µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
18718878
![]() |
R&D Systems
MAB3292-100 |
100 μg |
516.00€
100µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
18786818
![]() |
R&D Systems
MAB3292-500 |
500 μg |
2055.00€
500µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
M-CSFR/CD115 Monoclonal specifically detects M-CSFR/CD115 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifica
| M-CSFR/CD115 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF1R, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, MCSFR, M-CSFR, Proto-oncogene c-Fms | |
| CSF1R | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 61715 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P07333.2 | |
| 1436 | |
| Mouse myeloma cell line NS0-derived recombinant human M-CSF R/CD115 Ile20-Glu512 (Pro54Ala) Accession # P07333.2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human M-CSF R/CD115 in ELISAs. In ELISAs, no cross-react with recombinant human (rh) M-CSF, rhGM-CSF, rhPDGF R alpha , rhPDGF R beta , or rmM-CSF is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title