missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAEA Partial ORF (NP_001017405.1, 292 a.a. - 396 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010296-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172316

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene product mediates the attachment of erythroblasts to macrophages. This attachment promotes terminal maturation and enucleation of erythroblasts, presumably by suppressing apoptosis. This protein is an integral membrane protein with the N-terminus on the extracellular side and the C-terminus on the cytoplasmic side of the cell. Two immunologically related isoforms of erythroblast macrophage protein with apparent molecular weights of 33 kD and 36 kD were detected in macrophage membranes; this gene encodes the larger isoform. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined. [provided by RefSeq]

Sequence: AGLSAIKTPQCYKEDGSSKSPDCPVCSRSLNKLAQPLPMAHCANSRLVCKISGDVMNENNPPMMLPNGYVYGYNSLLSIRQDDKVVCPRTKEVFHFSQAEKVYIM
Specifica

Specifica

NP_001017405.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AGLSAIKTPQCYKEDGSSKSPDCPVCSRSLNKLAQPLPMAHCANSRLVCKISGDVMNENNPPMMLPNGYVYGYNSLLSIRQDDKVVCPRTKEVFHFSQAEKVYIM
RUO
MAEA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10296
MAEA (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EMLP/EMP/HLC-10/PIG5
MAEA
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali