Learn More
Abnova™ Human MARCH5 Full-length ORF (NP_060294.1, 1 a.a. - 278 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054708-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2 (MFN2; MIM 608507) and DRP1 (DNM1L; MIM 603850) (Nakamura et al., 2006 [PubMed 16936636]).[supplied by OMIM]
Sequence: MPDQALQQMLDRSCWVCFATDEDDRTAEWVRPCRCRGSTKWVHQACLQRWVDEKQRGNSTARVACPQCNAEYLIVFPKLGPVVYVLDLADRLISKACPFAAAGIMVGSIYWTAVTYGAVTVMQVVGHKEGLDVMERADPLFLLIGLPTIPVMLILGKMIRWEDYVLRLWRKYSNKLQILNSIFPGIGCPVPRIPAEANPLADHVSATRILCGALVFPTIATIVGKLMFSSVNSNLQRTILGGIAFVAIKGAFKVYFKQQQYLRQAHRKILNYPEQEEASpecifica
NP_060294.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20445/MARCH-V/MITOL/RNF153 | |
MARCH5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54708 | |
MARCH5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPDQALQQMLDRSCWVCFATDEDDRTAEWVRPCRCRGSTKWVHQACLQRWVDEKQRGNSTARVACPQCNAEYLIVFPKLGPVVYVLDLADRLISKACPFAAAGIMVGSIYWTAVTYGAVTVMQVVGHKEGLDVMERADPLFLLIGLPTIPVMLILGKMIRWEDYVLRLWRKYSNKLQILNSIFPGIGCPVPRIPAEANPLADHVSATRILCGALVFPTIATIVGKLMFSSVNSNLQRTILGGIAFVAIKGAFKVYFKQQQYLRQAHRKILNYPEQEEA | |
RUO | |
MARCH5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |