missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MARCH7 Partial ORF (NP_073737, 66 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00064844-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: WYSESEITQGARSRSQNQQRDHDSKRPKLSCTNCTTSAGRNVGNGLNTLSDSSWRHSQVPRSSSMVLGSFGEspecificaciones
NP_073737 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.55kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
WYSESEITQGARSRSQNQQRDHDSKRPKLSCTNCTTSAGRNVGNGLNTLSDSSWRHSQVPRSSSMVLGSFG | |
RUO | |
MARCH7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64844 | |
MARCH7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AXO/AXOT/DKFZp586F1122/MARCH-VII/RNF177 | |
MARCH7 | |
Recombinant | |
wheat germ expression system |