Learn More
Abnova™ Human MCC Full-length ORF (NP_002378.1, 1 a.a. - 829 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004163-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene is a candidate colorectal tumor suppressor gene that is thought to negatively regulate cell cycle progression. The orthologous gene in the mouse expresses a phosphoprotein associated with the plasma membrane and membrane organelles, and overexpression of the mouse protein inhibits entry into S phase. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MNSGVAMKYGNDSSAELSELHSAALASLKGDIVELNKRLQQTERERDLLEKKLAKAQCEQSHLMREHEDVQERTTLRYEERITELHSVIAELNKKIDRLQGTTIREEDEYSELRSELSQSQHEVNEDSRSMDQDQTSVSIPENQSTMVTADMDNCSDLNSELQRVLTGLENVVCGRKKSSCSLSVAEVDRHIEQLTTASEHCDLAIKTVEEIEGVLGRDLYPNLAEERSRWEKELAGLREENESLTAMLCSKEEELNRTKATMNAIREERDRLRRRVRELQTRLQSVQATGPSSPGRLTSTNRPINPSTGELSTSSSSNDIPIAKIAERVKLSKTRSESSSSDRPVLGSEISSIGVSSSVAEHLAHSLQDCSNIQEIFQTLYSHGSAISESKIREFEVETERLNSRIEHLKSQNDLLTITLEECKSNAERMSMLVGKYESNATALRLALQYSEQCIEAYELLLALAESEQSLILGQFRAAGVGSSPGDQSGDENITQMLKRAHDCRKTAENAAKALLMKLDGSCGGAFAVAGCSVQPWESLSSNSHTSTTSSTASSCDTEFTKEDEQRLKDYIQQLKNDRAAVKLTMLELESIHIDPLSYDVKPRGDSQRLDLENAVLMQELMAMKEEMAELKAQLYLLEKEKKALELKLSTREAQEQAYLVHIEHLKSEVEEQKEQRMRSLSSTSSGSKDKPGKECADAASPALSLAELRTTCSENELAAEFTNAIRREKKLKARVQELVSALERLTKSSEIRHQQSAEFVNDLKRANSNLVAAYEKAKKKHQNKLKKLESQMMAMVERHETQVRMLKQRIALLEEENSRPHTNETSLSpecifica
NP_002378.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
119.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp762O1615/FLJ38893/FLJ46755/MCC1 | |
MCC | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4163 | |
MCC (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNSGVAMKYGNDSSAELSELHSAALASLKGDIVELNKRLQQTERERDLLEKKLAKAQCEQSHLMREHEDVQERTTLRYEERITELHSVIAELNKKIDRLQGTTIREEDEYSELRSELSQSQHEVNEDSRSMDQDQTSVSIPENQSTMVTADMDNCSDLNSELQRVLTGLENVVCGRKKSSCSLSVAEVDRHIEQLTTASEHCDLAIKTVEEIEGVLGRDLYPNLAEERSRWEKELAGLREENESLTAMLCSKEEELNRTKATMNAIREERDRLRRRVRELQTRLQSVQATGPSSPGRLTSTNRPINPSTGELSTSSSSNDIPIAKIAERVKLSKTRSESSSSDRPVLGSEISSIGVSSSVAEHLAHSLQDCSNIQEIFQTLYSHGSAISESKIREFEVETERLNSRIEHLKSQNDLLTITLEECKSNAERMSMLVGKYESNATALRLALQYSEQCIEAYELLLALAESEQSLILGQFRAAGVGSSPGDQSGDENITQMLKRAHDCRKTAENAAKALLMKLDGSCGGAFAVAGCSVQPWESLSSNSHTSTTSSTASSCDTEFTKEDEQRLKDYIQQLKNDRAAVKLTMLELESIHIDPLSYDVKPRGDSQRLDLENAVLMQELMAMKEEMAELKAQLYLLEKEKKALELKLSTREAQEQAYLVHIEHLKSEVEEQKEQRMRSLSSTSSGSKDKPGKECADAASPALSLAELRTTCSENELAAEFTNAIRREKKLKARVQELVSALERLTKSSEIRHQQSAEFVNDLKRANSNLVAAYEKAKKKHQNKLKKLESQMMAMVERHETQVRMLKQRIALLEEENSRPHTNETSL | |
RUO | |
MCC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |