Learn More
Abnova™ Human MGEA5 Partial ORF (NP_036347, 823 a.a. - 916 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010724-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The dynamic modification of cytoplasmic and nuclear proteins by O-linked N-acetylglucosamine (O-GlcNAc) addition and removal on serine and threonine residues is catalyzed by OGT (MIM 300255), which adds O-GlcNAc, and MGEA5, a glycosidase that removes O-GlcNAc modifications (Gao et al., 2001 [PubMed 11148210]).[supplied by OMIM]
Sequence: SFHEEQEVLPETFLANFPSLIKMDIHKKVTDPSVAKSMMACLLSSLKANGSRGAFCEVRPDDKRILEFYSKLGCFEIAKMEGFPKDVVILGRSLSpecifica
NP_036347 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SFHEEQEVLPETFLANFPSLIKMDIHKKVTDPSVAKSMMACLLSSLKANGSRGAFCEVRPDDKRILEFYSKLGCFEIAKMEGFPKDVVILGRSL | |
RUO | |
MGEA5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10724 | |
MGEA5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11229/FLJ23355/KIAA0679/MEA5/NCOAT/OGA | |
MGEA5 | |
Recombinant | |
wheat germ expression system |