missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MMP10 Partial ORF (NP_002416.1, 271 a.a. - 368 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004319-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16131315

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades proteoglycans and fibronectin. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. [provided by RefSeq]

Sequence: EPLVPTKSVPSGSEMPAKCDPALSFDAISTLRGEYLFFKDRYFWRRSHWNPEPEFHLISAFWPSLPSYLDAAYEVNSRDTVFIFKGNEFWAIRGNEVQ
Specifica

Specifica

NP_002416.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EPLVPTKSVPSGSEMPAKCDPALSFDAISTLRGEYLFFKDRYFWRRSHWNPEPEFHLISAFWPSLPSYLDAAYEVNSRDTVFIFKGNEFWAIRGNEVQ
RUO
MMP10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4319
MMP10 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SL-2/STMY2
MMP10
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali