missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MTMR12 Partial ORF (NP_061934, 648 a.a. - 747 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00054545-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16127606

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Phosphatidylinositide 3-kinase-derived membrane-anchored phosphatidylinositides, such as phosphatidylinositol 3-phosphate (PtdIns(3)P), regulate diverse cellular processes. The protein encoded by this gene functions as an adaptor subunit in a complex with an active PtdIns(3)P 3-phosphatase. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined. [provided by RefSeq]

Sequence: PEAQILGGGQVATLSKLLEMMEEVQSLQEKIDERHHSQQAPQAEAPCLLRNSARLSSLFPFALLQRHSSKPVLPTSGWKALGDEDDLAKREDEFVDLGDV
Specifica

Specifica

NP_061934
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PEAQILGGGQVATLSKLLEMMEEVQSLQEKIDERHHSQQAPQAEAPCLLRNSARLSSLFPFALLQRHSSKPVLPTSGWKALGDEDDLAKREDEFVDLGDV
RUO
MTMR12
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
54545
MTMR12 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
3-PAP/PIP3AP
MTMR12
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali