Learn More
Abnova™ Human MYO5A Partial ORF (NP_000250.1, 1758 a.a. - 1853 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004644-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene is one of three myosin V heavy-chain genes, belonging to the myosin gene superfamily. Myosin V is a class of actin-based motor proteins involved in cytoplasmic vesicle transport and anchorage, spindle-pole alignment and mRNA translocation. The protein encoded by this gene is abundant in melanocytes and nerve cells. Mutations in this gene cause Griscelli syndrome type-1 (GS1), Griscelli syndrome type-3 (GS3) and neuroectodermal melanolysosomal disease, or Elejalde disease. Multiple alternatively spliced transcript variants encoding different isoforms have been reported, but the full-length nature of some variants has not been determined. [provided by RefSeq]
Sequence: KKTDDDAEAICSMCNALTTAQIVKVLNLYTPVNEFEERVSVSFIRTIQMRLRDRKDSPQLLMDAKHIFPVTFPFNPSSLALETIQIPASLGLGFISSpecifica
NP_000250.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KKTDDDAEAICSMCNALTTAQIVKVLNLYTPVNEFEERVSVSFIRTIQMRLRDRKDSPQLLMDAKHIFPVTFPFNPSSLALETIQIPASLGLGFIS | |
RUO | |
MYO5A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4644 | |
MYO5A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GS1/MYH12/MYO5/MYR12 | |
MYO5A | |
Recombinant | |
wheat germ expression system |