missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEK9 Full-length ORF (AAH93881.1, 1 a.a. - 979 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00091754-P01.25ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Sequence: MSVLGEYERHCDSINSDFGSESGGCGDSSPGPSASQGPRAGGGAAEQEELHYIPIRVLGRGAFGEATLYRRTEDDSLVVWKEVDLTRLSEKERRDALNEIVILALLQHDNIIAYYNHFMDNTTLLIELEYCNGGNLYDKILRQKDKLFEEEMVVWYLFQIVSAVSCIHKAGILHRDIKTLNIFLTKANLIKLGDYGLAKKLNSEYSMAETLVGTPYYMSPELCQGVKYNFKSDIWAVGCVIFELLTLKRTFDATNPLNLCVKIVQGIRAMEVDSSQYSLELIQMVHSCLDQDPEQRPTADELLDRPLLRKRRREMEEKVTLLNAPTKRPRSSTVTEAPIAVVTSRTSEVYVWGGGKSTPQKLDVIKSGCSARQVCAGNTHFAVVTVEKELYTWVNMQGGTKLHGQLGHGDKASYRQPKHVEKLQGKAIRQVSCGDDFTVCVTDEGQLYAFGSDYYGCMGVDKVAGPEVLEPMQLNFFLSNPVEQVSCGDNHVVVLTRNKEVYSWGCGEYGRLGLDSEEDYYTPQKVDVPKALIIVAVQCGCDGTFLLTQSGKVLACGLNEFNKLGLNQCMSGIINHEAYHEVPYTTSFTLAKQLSFYKIRTIAPGKTHTAAIDERGRLLTFGCNKCGQLGVGNYKKRLGINLLGGPLGGKQVIRVSCGDEFTIAATDDNHIFAWGNGGNGRLAMTPTERPHGSDICTSWPRPIFGSLHHVPDLSCRGWHTILIVEKVLNSKTIRSNSSGLSIGTVFQSSSPGGGGGGGGGEEEDSQQESETPDPSGGFRGTMEADRGMEGLISPTEAMGNSNGASSSCPGWLRKELENAEFIPMPDSPSPLSAAFSESEKDTLPYEELQGLKVASEAPLEHKPQVEASSPRLNPAVTCAGKGTPLTPPACACSSLQVEVERLQGLVLKCLAEQQKLQQENLQIFTQLQKLNKKLEGGQQVGMHSKGTQTAKEEMEMDPKPDLDSDSWCLLGTDSCRPSLSpecifica
AAH93881.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
133.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 μg | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434D0935/MGC138306/MGC16714/NERCC/NERCC1/Nek8 | |
NEK9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
91754 | |
NEK9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSVLGEYERHCDSINSDFGSESGGCGDSSPGPSASQGPRAGGGAAEQEELHYIPIRVLGRGAFGEATLYRRTEDDSLVVWKEVDLTRLSEKERRDALNEIVILALLQHDNIIAYYNHFMDNTTLLIELEYCNGGNLYDKILRQKDKLFEEEMVVWYLFQIVSAVSCIHKAGILHRDIKTLNIFLTKANLIKLGDYGLAKKLNSEYSMAETLVGTPYYMSPELCQGVKYNFKSDIWAVGCVIFELLTLKRTFDATNPLNLCVKIVQGIRAMEVDSSQYSLELIQMVHSCLDQDPEQRPTADELLDRPLLRKRRREMEEKVTLLNAPTKRPRSSTVTEAPIAVVTSRTSEVYVWGGGKSTPQKLDVIKSGCSARQVCAGNTHFAVVTVEKELYTWVNMQGGTKLHGQLGHGDKASYRQPKHVEKLQGKAIRQVSCGDDFTVCVTDEGQLYAFGSDYYGCMGVDKVAGPEVLEPMQLNFFLSNPVEQVSCGDNHVVVLTRNKEVYSWGCGEYGRLGLDSEEDYYTPQKVDVPKALIIVAVQCGCDGTFLLTQSGKVLACGLNEFNKLGLNQCMSGIINHEAYHEVPYTTSFTLAKQLSFYKIRTIAPGKTHTAAIDERGRLLTFGCNKCGQLGVGNYKKRLGINLLGGPLGGKQVIRVSCGDEFTIAATDDNHIFAWGNGGNGRLAMTPTERPHGSDICTSWPRPIFGSLHHVPDLSCRGWHTILIVEKVLNSKTIRSNSSGLSIGTVFQSSSPGGGGGGGGGEEEDSQQESETPDPSGGFRGTMEADRGMEGLISPTEAMGNSNGASSSCPGWLRKELENAEFIPMPDSPSPLSAAFSESEKDTLPYEELQGLKVASEAPLEHKPQVEASSPRLNPAVTCAGKGTPLTPPACACSSLQVEVERLQGLVLKCLAEQQKLQQENLQIFTQLQKLNKKLEGGQQVGMHSKGTQTAKEEMEMDPKPDLDSDSWCLLGTDSCRPSL | |
RUO | |
NEK9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto