missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NID2 Full-length ORF (AAH35608.1, 1 a.a. - 969 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00022795-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16126592

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Basement membranes, which are composed of type IV collagens (see MIM 120130), laminins (see LAMC1; MIM 150290), perlecan (HSPG2; MIM 142461), and nidogen (see NID1; MIM 131390), are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.[supplied by OMIM]

Sequence: MEGDRVAGRPVLSSLPVLLLLQLLMLRAAALHPDELFPHGESWGDQLLQEGDDESSAVVKLANPLHFYEARFSNLYVGTNGIISTQDFPRETQYVDYDFPTDFPAIAPFLADIDTSHGRGRVLYRVNGKVSGHLHVGHTPVHFTDVDLHAYIVGNDGRAYTAISHIPQPAAQALLPLTPIGGLFGWLFALEKPGSENGFSLAGAAFTHDMEVTFYPGEETVRITQTAEGLDPENYLSIKTNIQGQVPYVPANFTAHISPYKELYHYSDSTVTSTSSRDYSLTFGAINQTWSYRIHQNITYQVCRHAPRHPSFPTTQQLNVDRVFALYNDEERVLRFAVTNQIGPVKEDSDPTPVNPCYDGSHMCDTTARCHPGTGVDYTCECASGYQGDGRNCVDENECATGFHRCGPNSVCINLPGSYRCECRSAYEFADDRHTCILITPPANPCEDGSHTCAPAGQARCVHHGGSTFSCACLPGYAGDGHQCTDVDECSENRCHPAATCYNTPGSFSCRCQPGYYGDGFQCIPDSTSSLTPCEQQQRHAQAQYAYPGARFHIPQCDEQGNFLPLQCHGSTGFCWCVDPDGHEVPGTQTPPGSTPPHCGPSPEPTQRPPTICERWRENLLEHYGGTPRDDQYVPQCDDLGHFIPLQCHGKSDFCWCVDKDGREVQGTRSQPGTTPACIPTVAPPMVRPTPRPDVTPPSVGTFLLYTQGQQIGYLPLNGTRLQKDAAKTLLSLHGSIIVGIDYDCRERMVYWTDVAGRTISRAGLELGAEPETIVNSGLISPEGLAIDHIRRTMYWTDSVLDKIESALLDGSERKVLFYTDLVNPRAIAVDPIRGNLYWTDWNREAPKIETSSLDGENRRILINTDIGLPNGLTFDPFSKLLCWADAGTKKLECTLPDGTGRRVIQNNLKYPFSIVSYADHFYHTDWRRDGVVSVNKHSGQFTDEYLPEQRSHLYGITAVYPYCPTGRK
Specifica

Specifica

AAH35608.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
133.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NID2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
22795
NID2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEGDRVAGRPVLSSLPVLLLLQLLMLRAAALHPDELFPHGESWGDQLLQEGDDESSAVVKLANPLHFYEARFSNLYVGTNGIISTQDFPRETQYVDYDFPTDFPAIAPFLADIDTSHGRGRVLYRVNGKVSGHLHVGHTPVHFTDVDLHAYIVGNDGRAYTAISHIPQPAAQALLPLTPIGGLFGWLFALEKPGSENGFSLAGAAFTHDMEVTFYPGEETVRITQTAEGLDPENYLSIKTNIQGQVPYVPANFTAHISPYKELYHYSDSTVTSTSSRDYSLTFGAINQTWSYRIHQNITYQVCRHAPRHPSFPTTQQLNVDRVFALYNDEERVLRFAVTNQIGPVKEDSDPTPVNPCYDGSHMCDTTARCHPGTGVDYTCECASGYQGDGRNCVDENECATGFHRCGPNSVCINLPGSYRCECRSAYEFADDRHTCILITPPANPCEDGSHTCAPAGQARCVHHGGSTFSCACLPGYAGDGHQCTDVDECSENRCHPAATCYNTPGSFSCRCQPGYYGDGFQCIPDSTSSLTPCEQQQRHAQAQYAYPGARFHIPQCDEQGNFLPLQCHGSTGFCWCVDPDGHEVPGTQTPPGSTPPHCGPSPEPTQRPPTICERWRENLLEHYGGTPRDDQYVPQCDDLGHFIPLQCHGKSDFCWCVDKDGREVQGTRSQPGTTPACIPTVAPPMVRPTPRPDVTPPSVGTFLLYTQGQQIGYLPLNGTRLQKDAAKTLLSLHGSIIVGIDYDCRERMVYWTDVAGRTISRAGLELGAEPETIVNSGLISPEGLAIDHIRRTMYWTDSVLDKIESALLDGSERKVLFYTDLVNPRAIAVDPIRGNLYWTDWNREAPKIETSSLDGENRRILINTDIGLPNGLTFDPFSKLLCWADAGTKKLECTLPDGTGRRVIQNNLKYPFSIVSYADHFYHTDWRRDGVVSVNKHSGQFTDEYLPEQRSHLYGITAVYPYCPTGRK
RUO
NID2
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali