Learn More
ACROBiosystems Human NOTCH1 Protein, Fc Tag
MALS verified
Marca:  ACROBiosystems NO1-H5255-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17106551
  • 453.47€ / 100µg
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
NOTCH1 Interacts with DNER, DTX1, DTX2 and RBPJ/RBPSUH. Also interacts with MAML1, MAML2 and MAML3 which act as transcriptional coactivators for NOTCH1. The NOTCH1 intracellular domain interacts with SNW1; the interaction involves multimerized NOTCH1 NICD and is implicated in a formation of an intermediate preactivation complex which associates with DNA-bound CBF-1/RBPJ. The activated membrane-bound form interacts with AAK1 which promotes NOTCH1 stabilization. Functions as a receptor for membrane-bound ligands Jagged-1 (JAG1), Jagged-2 (JAG2) and Delta-1 (DLL1) to regulate cell-fate determination. Involved in the maturation of both CD4+ and CD8+ cells in the thymus. Important for follicular differentiation and possibly cell fate selection within the follicle. During cerebellar development, functions as a receptor for neuronal DNER and is involved in the differentiation of Bergmann glia.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 80.0 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
NP_060087.3
100 μg
Ala 19 - Gln 526
NOTCH1,Notch 1,hN1,TAN1
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
80.0 kDa
Human NOTCH1, Fc Tag (NO1-H5255) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Gln 526 (Accession # P46531-1).
-20°C
NOTCH1
Human
PBS, pH7.4
HEK293
Other Recombinant Proteins