missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NRN1 Full-length ORF (NP_057672.1, 1 a.a. - 142 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00051299-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129882

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene is expressed in postmitotic-differentiating neurons of the developmental nervous system and neuronal structures associated with plasticity in the adult. The expression of this gene can be induced by neural activity and neurotrophins. The encoded protein contains a consensus cleavage signal found in glycosylphoshatidylinositol (GPI)-anchored proteins. In vitro assays demonstrated that this protein promotes neurite outgrowth and arborization, suggesting its role in promoting neuritogenesis. [provided by RefSeq]

Sequence: MGLKLNGRYISLILAVQIAYLVQAVRAAGKCDAVFKGFSDCLLKLGDSMANYPQGLDDKTNIKTVCTYWEDFHSCTVTALTDCQEGAKDMWDKLRKESKNLNIQGSLFELCGSGNGAAGSLLPAFPVLLVSLSAALATWLSF
Specifica

Specifica

NP_057672.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
41.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC44811/NRN/dJ380B8.2
NRN1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51299
NRN1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGLKLNGRYISLILAVQIAYLVQAVRAAGKCDAVFKGFSDCLLKLGDSMANYPQGLDDKTNIKTVCTYWEDFHSCTVTALTDCQEGAKDMWDKLRKESKNLNIQGSLFELCGSGNGAAGSLLPAFPVLLVSLSAALATWLSF
RUO
NRN1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali