missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NTNG2 Partial ORF (NP_115925, 21 a.a. - 119 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084628-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: ICKSWVTTDEGPTWEFYACQPKVMRLKDYVKVKVEPSGITCGDPPERFCSHENPYLCSNECDASNPDLAHPPRLMFDKEEEGLATYWQSITWSRYPSPLSpecifica
NP_115925 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ICKSWVTTDEGPTWEFYACQPKVMRLKDYVKVKVEPSGITCGDPPERFCSHENPYLCSNECDASNPDLAHPPRLMFDKEEEGLATYWQSITWSRYPSPL | |
RUO | |
NTNG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84628 | |
NTNG2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0625/KIAA1857/LHLL9381/Lmnt2/MGC21884/NTNG1/bA479K20.1 | |
NTNG2 | |
Recombinant | |
wheat germ expression system |