missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR6N1 Full-length ORF (NP_001005185.1, 1 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00128372-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16127813

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: MDTGNWSQVAEFIILGFPHLQGVQIYLFLLLLLIYLMTVLGNLLIFLVVCLDSRLHTPMYHFVSILSFSELGYTAATIPKMLANLLSEKKTISFSGCLLQIYFFHSLGATECYLLTAMAYDRYLAICRPLHYPTLMTPTLCAEIAIGCWLGGLAGPVVEISLISRLPFCGPNRIQHVFCDFPPVLSLACTDTSINVLVDFVINSCKILATFLLILCSYVQIICTVLRIPSAAGKRKAISTCASHFTVVLIFYGSILSMYVQLKKSYSLDYDQALAVVYSVLTPFLNPFIYSLRNKEIKEAVRRQLKRIGILA
Specifica

Specifica

NP_001005185.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OR1-22
OR6N1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
128372
OR6N1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDTGNWSQVAEFIILGFPHLQGVQIYLFLLLLLIYLMTVLGNLLIFLVVCLDSRLHTPMYHFVSILSFSELGYTAATIPKMLANLLSEKKTISFSGCLLQIYFFHSLGATECYLLTAMAYDRYLAICRPLHYPTLMTPTLCAEIAIGCWLGGLAGPVVEISLISRLPFCGPNRIQHVFCDFPPVLSLACTDTSINVLVDFVINSCKILATFLLILCSYVQIICTVLRIPSAAGKRKAISTCASHFTVVLIFYGSILSMYVQLKKSYSLDYDQALAVVYSVLTPFLNPFIYSLRNKEIKEAVRRQLKRIGILA
RUO
OR6N1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali