missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OSBP Partial ORF (NP_002547.1, 324 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005007-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132245

  • 528.00€ / 25µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Oxysterol binding protein is an intracellular protein that is believed to transport sterols from lysosomes to the nucleus where the sterol down-regulates the genes for the LDL receptor, HMG-CoA reductase, and HMG synthetase [provided by RefSeq]

Sequence: GATVLPANTPGNVGSGKDQCCSGKGDMSDEDDENEFFDAPEIITMPENLGHKRTGSNISGASSDISLDEQYKHQLEETKKEKRT
Specifica

Specifica

NP_002547.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.98kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GATVLPANTPGNVGSGKDQCCSGKGDMSDEDDENEFFDAPEIITMPENLGHKRTGSNISGASSDISLDEQYKHQLEETKKEKRT
RUO
OSBP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5007
OSBP (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OSBP1
OSBP
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali