missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OSBPL11 Full-length ORF (NP_073613.2, 1 a.a. - 747 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00114885-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16127153

  • 349.00€ / 10µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. [provided by RefSeq]

Sequence: MQGGEPVSTMKVSESEGKLEGQATAVTPNKNSSCGGGISSSSSSRGGSAKGWQYSDHMENVYGYLMKYTNLVTGWQYRFFVLNNEAGLLEYFVNEQSRNQKPRGTLQLAGAVISPSDEDSHTFTVNAASGEQYKLRATDAKERQHWVSRLQICTQHHTEAIGKNNPPLKSRSFSLASSSNSPISQRRPSQNAISFFNVGHSKLQSLSKRTNLPPDHLVEVREMMSHAEGQQRDLIRRIECLPTSGHLSSLDQDLLMLKATSMATMNCLNDCFHILQLQHASHQKGSLPSGTTIEWLEPKISLSNHYKNGADQPFATDQSKPVAVPEEQPVAESGLLAREPEEINADDEIEDTCDHKEDDLGAVEEQRSVILHLLSQLKLGMDLTRVVLPTFILEKRSLLEMYADFMSHPDLFIAITNGATAEDRMIRFVEYYLTSFHEGRKGAIAKKPYNPIIGETFHCSWKMPKSEVASSVFSSSSTQGVTNHAPLSGESLTQVGSDCYTVRFVAEQVSHHPPVSGFYAECTERKMCVNAHVWTKSKFLGMSIGVTMVGEGILSLLEHGEEYTFSLPCAYARSILTVPWVELGGKVSVNCAKTGYSASITFHTKPFYGGKLHRVTAEVKHNITNTVVCRVQGEWNSVLEFTYSNGETKYVDLTKLAVTKKRVRPLEKQDPFESRRLWKNVTDSLRESEIDKATEHKHTLEERQRTEERHRTETGTPWKTKYFIKEGDGWVYHKPLWKIIPTTQPAE
Specifica

Specifica

NP_073613.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
110kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ13012/FLJ13164/ORP-11/ORP11/OSBP12/TCCCIA00292
OSBPL11
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
114885
OSBPL11 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQGGEPVSTMKVSESEGKLEGQATAVTPNKNSSCGGGISSSSSSRGGSAKGWQYSDHMENVYGYLMKYTNLVTGWQYRFFVLNNEAGLLEYFVNEQSRNQKPRGTLQLAGAVISPSDEDSHTFTVNAASGEQYKLRATDAKERQHWVSRLQICTQHHTEAIGKNNPPLKSRSFSLASSSNSPISQRRPSQNAISFFNVGHSKLQSLSKRTNLPPDHLVEVREMMSHAEGQQRDLIRRIECLPTSGHLSSLDQDLLMLKATSMATMNCLNDCFHILQLQHASHQKGSLPSGTTIEWLEPKISLSNHYKNGADQPFATDQSKPVAVPEEQPVAESGLLAREPEEINADDEIEDTCDHKEDDLGAVEEQRSVILHLLSQLKLGMDLTRVVLPTFILEKRSLLEMYADFMSHPDLFIAITNGATAEDRMIRFVEYYLTSFHEGRKGAIAKKPYNPIIGETFHCSWKMPKSEVASSVFSSSSTQGVTNHAPLSGESLTQVGSDCYTVRFVAEQVSHHPPVSGFYAECTERKMCVNAHVWTKSKFLGMSIGVTMVGEGILSLLEHGEEYTFSLPCAYARSILTVPWVELGGKVSVNCAKTGYSASITFHTKPFYGGKLHRVTAEVKHNITNTVVCRVQGEWNSVLEFTYSNGETKYVDLTKLAVTKKRVRPLEKQDPFESRRLWKNVTDSLRESEIDKATEHKHTLEERQRTEERHRTETGTPWKTKYFIKEGDGWVYHKPLWKIIPTTQPAE
RUO
OSBPL11
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali