missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OTOA Partial ORF (NP_653273.3, 27 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00146183-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132037

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is specifically expressed in the inner ear, and is located at the interface between the apical surface of the inner ear sensory epithelia and their overlying acellular gels. It is prposed that this protein is involved in the attachment of the inner ear acellular gels to the apical surface of the underlying nonsensory cells. Mutations in this gene are associated with autosomal recessive deafness type 22 (DFNB22). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: NSRQDLHPLLQNMAEEIIDGSYLNALLDLIQFQSSHVWTDDLSHRVLAYLNSRNVAFTIPSLQAAVENHLEQRLHQPQKLLEDLRKTDAQQFRTAMKC
Specifica

Specifica

NP_653273.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NSRQDLHPLLQNMAEEIIDGSYLNALLDLIQFQSSHVWTDDLSHRVLAYLNSRNVAFTIPSLQAAVENHLEQRLHQPQKLLEDLRKTDAQQFRTAMKC
RUO
OTOA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
146183
OTOA (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DFNB22/FLJ32773/MGC157747/MGC39813
OTOA
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali