missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PBX2 Partial ORF (NP_002577, 354 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005089-Q02.25ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6. [provided by RefSeq]
Sequence: DMFLGMPGLNGDSYSASQVESLRHSMGPGGYGDNLGGGQMYSPREMRANGSWQEAVTPSSVTSPTEGPGSVHSDTSNSpecifica
NP_002577 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.21kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DMFLGMPGLNGDSYSASQVESLRHSMGPGGYGDNLGGGQMYSPREMRANGSWQEAVTPSSVTSPTEGPGSVHSDTSN | |
RUO | |
PBX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5089 | |
PBX2 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
G17/HOX12/PBX2MHC | |
PBX2 | |
Recombinant | |
wheat germ expression system |