missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCDHB15 Full-length ORF (AAH38797, 1 a.a. - 787 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00056121-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172073

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]

Sequence: MEPAGERFPEQRQVLILLLLLEVTLAGWEPRRYSVMEETERGSFVANLANDLGLGVGELAERGARVVSEDNEQGLQLDLQTGQLILNEKLDREKLCGPTEPCIMHFQVLLKKPLEVFRAELLVTDINDHSPEFPEREMTLKIPETSSLGTVFPLKKARDLDVGSNNVQNYNISPNSHFHVSTRTRGDGRKYPELVLDTELDREEQAELRLTLTAVDGGSPPRSGTVQILILVLDANDNAPEFVQALYEVQVPENSPVGSLVVKVSARDLDTGTNGEISYSLYYSSQEIDKPFELSSLSGEIRLIKKLDFETMSSYDLDIEASDGGGLSGKCSVSVKVLDVNDNFPELSISSLTSPIPENSPETEVALFRIRDRDSGENGKMICSIQDDVPFKLKPSVENFYRLVTEGALDRETRAEYNITITITDLGTPRLKTEQSITVLVSDVNDNAPAFTQTSYTLFVRENNSPALHIGSVRATDRDSGTNAQVTYSLLPPQDPHLPLTSLVSINTDNGHLFALQSLDYEALQAFEFRVGATDRGFPALSSEALVRVLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDVAKHRLVVLVKDNGEPPRSATATLQVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVFLFVAVRLCRRSRAASVGRCSVPEGPFPGHLVDVSGTGTLSQSYQYEVCLTGGSESNDFKFLKPIFPNIVSQDSRRKSEFLE
Specifica

Specifica

AAH38797
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
112.31kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PCDH-BETA15
PCDHB15
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
56121
PCDHB15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEPAGERFPEQRQVLILLLLLEVTLAGWEPRRYSVMEETERGSFVANLANDLGLGVGELAERGARVVSEDNEQGLQLDLQTGQLILNEKLDREKLCGPTEPCIMHFQVLLKKPLEVFRAELLVTDINDHSPEFPEREMTLKIPETSSLGTVFPLKKARDLDVGSNNVQNYNISPNSHFHVSTRTRGDGRKYPELVLDTELDREEQAELRLTLTAVDGGSPPRSGTVQILILVLDANDNAPEFVQALYEVQVPENSPVGSLVVKVSARDLDTGTNGEISYSLYYSSQEIDKPFELSSLSGEIRLIKKLDFETMSSYDLDIEASDGGGLSGKCSVSVKVLDVNDNFPELSISSLTSPIPENSPETEVALFRIRDRDSGENGKMICSIQDDVPFKLKPSVENFYRLVTEGALDRETRAEYNITITITDLGTPRLKTEQSITVLVSDVNDNAPAFTQTSYTLFVRENNSPALHIGSVRATDRDSGTNAQVTYSLLPPQDPHLPLTSLVSINTDNGHLFALQSLDYEALQAFEFRVGATDRGFPALSSEALVRVLVLDANDNSPFVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDVAKHRLVVLVKDNGEPPRSATATLQVLLVDGFSQPYLPLPEAAPAQAQADSLTVYLVVALASVSSLFLFSVFLFVAVRLCRRSRAASVGRCSVPEGPFPGHLVDVSGTGTLSQSYQYEVCLTGGSESNDFKFLKPIFPNIVSQDSRRKSEFLE
RUO
PCDHB15
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali