Learn More
Abnova™ Human PFKM Partial ORF (NP_000280, 681 a.a. - 780 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005213-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The PFKM gene encodes the muscle isoform of phosphofructokinase (PFK) (ATP:D-fructose-6-phosphate-1-phosphotransferase, EC 2.7.1.11). PFK catalyzes the irreversible conversion of fructose-6-phosphate to fructose-1,6-bisphosphate and is a key regulatory enzyme in glycolysis. Mammalian PFK is a tetramer made up of various combinations of 3 subunits: muscle (PFKM), liver (PFKL; MIM 171860), and platelet (PFKP; MIM 171840), the genes for which are located on chromosomes 12q13, 21q22, and 10p, respectively. The composition of the tetramers differs according to the tissue type. Muscle and liver PFK are a homotetramers of 4M and 4L subunits, respectively. Erythrocytes contain both L and M subunits, which randomly tetramerize to form M4, L4, and M3L, M2L2, and ML3 hybrid forms of the holoenzyme (Vora et al., 1980 [PubMed 6444721]; Raben and Sherman, 1995 [PubMed 7550225]).[supplied by OMIM]
Sequence: AKAMNWMSGKIKESYRNGRIFANTPDSGCVLGMRKRALVFQPVAELKDQTDFEHRIPKEQWWLKLRPILKILAKYEIDLDTSDHAHLEHITRKRSGEAAVSpecifica
NP_000280 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AKAMNWMSGKIKESYRNGRIFANTPDSGCVLGMRKRALVFQPVAELKDQTDFEHRIPKEQWWLKLRPILKILAKYEIDLDTSDHAHLEHITRKRSGEAAV | |
RUO | |
PFKM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5213 | |
PFKM (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GSD7/MGC8699/PFK-1/PFK-M/PFKX | |
PFKM | |
Recombinant | |
wheat germ expression system |