Learn More
Abnova™ Human PIGM Partial ORF (NP_660150, 35 a.a. - 96 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00093183-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a transmembrane protein that is located in the endoplasmic reticulum and is involved in GPI-anchor biosynthesis. The glycosylphosphatidylinositol (GPI)-anchor is a glycolipid which contains three mannose molecules in its core backbone. The GPI-anchor is found on many blood cells and serves to anchor proteins to the cell surface. This gene encodes a mannosyltransferase, GPI-MT-I, that transfers the first mannose to GPI on the lumenal side of the endoplasmic reticulum. [provided by RefSeq]
Sequence: YGVFQDRTLHVRYTDIDYQVFTDAARFVTEGRSPYLRATYRYTPLLGWLLTPNIYLSELFGKSpecifica
NP_660150 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.56kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YGVFQDRTLHVRYTDIDYQVFTDAARFVTEGRSPYLRATYRYTPLLGWLLTPNIYLSELFGK | |
RUO | |
PIGM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
93183 | |
PIGM (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPI-MT-I/MGC29896 | |
PIGM | |
Recombinant | |
wheat germ expression system |