Learn More
Abnova™ Human PLA2G6 Full-length ORF (NP_001004426.1, 1 a.a. - 752 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008398-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is an A2 phospholipase, a class of enzyme that catalyzes the release of fatty acids from phospholipids. The encoded protein may play a role in phospholipid remodelling, arachidonic acid release, leukotriene and prostaglandin synthesis, fas-mediated apoptosis, and transmembrane ion flux in glucose-stimulated B-cells. Several transcript variants encoding multiple isoforms have been described, but the full-length nature of only two of them have been determined to date. [provided by RefSeq]
Sequence: MQFFGRLVNTFSGVTNLFSNPFRVKEVAVADYTSSDRVREEGQLILFQNTPNRTWDCVLVNPRNSQSGFRLFQLELEADALVNFHQYSSQLLPFYESSPQVLHTEVLQHLTDLIRNHPSWSVAHLAVELGIRECFHHSRIISCANCAENEEGCTPLHLACRKGDGEILVELVQYCHTQMDVTDYKGETVFHYAVQGDNSQVLQLLGRNAVAGLNQVNNQGLTPLHLACQLGKQEMVRVLLLCNARCNIMGPNGYPIHSAMKFSQKGCAEMIISMDSSQIHSKDPRYGASPLHWAKNAEMARMLLKRGCNVNSTSSAGNTALHVAVMRNRFDCAIVLLTHGANADARGEHGNTPLHLAMSKDNVEMIKALIVFGAEVDTPNDFGETPTFLASKIGRQLQDLMHISRARKPAFILGSMRDEKRTHDHLLCLDGGGVKGLIIIQLLIAIEKASGVATKDLFDWVAGTSTGGILALAILHSKSMAYMRGMYFRMKDEVFRGSRPYESGPLEEFLKREFGEHTKMTDVRKPKVMLTGTLSDRQPAELHLFRNYDAPETVREPRFNQNVNLRPPAQPSDQLVWRAARSSGAAPTYFRPNGRFLDGGLLANNPTLDAMTEIHEYNQDLIRKGQANKVKKLSIVVSLGTGRSPQVPVTCVDVFRPSNPWELAKTVFGAKELGKMVVDCCTDPDGRAVDRARAWCEMVGIQYFRLNPQLGTDIMLDEVSDTVLVNALWETEVYIYEHREEFQKLIQLLLSPSpecifica
NP_001004426.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CaI-PLA2/GVI/INAD1/IPLA2-VIA/PARK14/PLA2/PNPLA9/iPLA2 | |
PLA2G6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8398 | |
PLA2G6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQFFGRLVNTFSGVTNLFSNPFRVKEVAVADYTSSDRVREEGQLILFQNTPNRTWDCVLVNPRNSQSGFRLFQLELEADALVNFHQYSSQLLPFYESSPQVLHTEVLQHLTDLIRNHPSWSVAHLAVELGIRECFHHSRIISCANCAENEEGCTPLHLACRKGDGEILVELVQYCHTQMDVTDYKGETVFHYAVQGDNSQVLQLLGRNAVAGLNQVNNQGLTPLHLACQLGKQEMVRVLLLCNARCNIMGPNGYPIHSAMKFSQKGCAEMIISMDSSQIHSKDPRYGASPLHWAKNAEMARMLLKRGCNVNSTSSAGNTALHVAVMRNRFDCAIVLLTHGANADARGEHGNTPLHLAMSKDNVEMIKALIVFGAEVDTPNDFGETPTFLASKIGRQLQDLMHISRARKPAFILGSMRDEKRTHDHLLCLDGGGVKGLIIIQLLIAIEKASGVATKDLFDWVAGTSTGGILALAILHSKSMAYMRGMYFRMKDEVFRGSRPYESGPLEEFLKREFGEHTKMTDVRKPKVMLTGTLSDRQPAELHLFRNYDAPETVREPRFNQNVNLRPPAQPSDQLVWRAARSSGAAPTYFRPNGRFLDGGLLANNPTLDAMTEIHEYNQDLIRKGQANKVKKLSIVVSLGTGRSPQVPVTCVDVFRPSNPWELAKTVFGAKELGKMVVDCCTDPDGRAVDRARAWCEMVGIQYFRLNPQLGTDIMLDEVSDTVLVNALWETEVYIYEHREEFQKLIQLLLSP | |
RUO | |
PLA2G6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |