Learn More
Abnova™ Human PLAGL2 Partial ORF (NP_002648, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00005326-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16129161
  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile
Descrizione
Descrizione
Pleiomorphic adenoma gene-like 2 is a zinc-finger protein that recognizes DNA and/or RNA. [provided by RefSeq]
Sequence: MTTFFTSVPPWIQDAKQEEEVGWKLVPRPRGREAESQVKCQCEISGTPFSNGEKLRPHSLPQPEQRPYSCPQLHCGKAFASKYKLYRHMATHSAQKPHQC
Specifica
Specifica
NP_002648
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MTTFFTSVPPWIQDAKQEEEVGWKLVPRPRGREAESQVKCQCEISGTPFSNGEKLRPHSLPQPEQRPYSCPQLHCGKAFASKYKLYRHMATHSAQKPHQC
RUO
PLAGL2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5326
PLAGL2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ23283
PLAGL2
Recombinant
wheat germ expression system