Learn More
Abnova™ Human PLCD3 Partial ORF (NP_588614, 514 a.a. - 613 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00113026-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the phospholipase C family, which catalyze the hydrolysis of phosphatidylinositol 4,5-bisphosphate to generate the second messengers diacylglycerol and inositol 1,4,5-trisphosphate (IP3). Diacylglycerol and IP3 mediate a variety of cellular responses to extracellular stimuli by inducing protein kinase C and increasing cytosolic Ca(2+) concentrations. This enzyme localizes to the plasma membrane and requires calcium for activation. Its activity is inhibited by spermine, sphingosine, and several phospholipids. [provided by RefSeq]
Sequence: AAAQRRLAKQISPELSALAVYCHATRLRTLHPAPNAPQPCQVSSLSERKAKKLIREAGNSFVRHNARQLTRVYPLGLRMNSANYSPQEMWNSGCQLVALNSpecifica
NP_588614 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AAAQRRLAKQISPELSALAVYCHATRLRTLHPAPNAPQPCQVSSLSERKAKKLIREAGNSFVRHNARQLTRVYPLGLRMNSANYSPQEMWNSGCQLVALN | |
RUO | |
PLCD3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
113026 | |
PLCD3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC71172 | |
PLCD3 | |
Recombinant | |
wheat germ expression system |