missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human POGK Full-length ORF (NP_060012.3, 1 a.a. - 609 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00057645-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132863

  • 528.00€ / 25µg
Spedizione stimata: 06-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The exact function of the protein encoded by this gene is not known. However, this gene product contains a KRAB domain (which is involved in protein-protein interactions) at the N-terminus, and a transposase domain at the C-terminus, suggesting that it may belong to the family of DNA-mediated transposons in human. [provided by RefSeq]

Sequence: MESTAYPLNLSLKEEEEEEEIQSRELEDGPADMQKVRICSEGGWVPALFDEVAIYFSDEEWEVLTEQQKALYREVMRMNYETVLSLEFPFPKPDMITRLEGEEESQNSDEWQLQGGTSAENEESDVKPPDWPNPMNATSQFPQPQHFDSFGLRLPRDITELPEWSEGYPFYMAMGFPGYDLSADDIAGKFQFSRGMRRSYDAGFKLMVVEYAESTNNCQAAKQFGVLEKNVRDWRKVKPQLQNAHAMRRAFRGPKNGRFALVDQRVAEYVRYMQAKGDPITREAMQLKALEIAQEMNIPEKGFKASLGWCRRMMRRYDLSLRHKVPVPQHLPEDLTEKLVTYQRSVLALRRAHDYEVAQMGNADETPICLEVPSRVTVDNQGEKPVLVKTPGREKLKITAMLGVLADGRKLPPYIILRGTYIPPGKFPSGMEIRCHRYGWMTEDLMQDWLEVVWRRRTGAVPKQRGMLILNGFRGHATDSVKNSMESMNTDMVIIPGGLTSQLQVLDVVVYKPLNDSVRAQYSNWLLAGNLALSPTGNAKKPPLGLFLEWVMVAWNSISSESIVQGFKKCHISSNLEEEDDVLWEIESELPGGGEPPKDCDTESMAESN
Specifica

Specifica

NP_060012.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
95.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BASS2/KIAA1513/KIAA15131/LST003
POGK
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
57645
POGK (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MESTAYPLNLSLKEEEEEEEIQSRELEDGPADMQKVRICSEGGWVPALFDEVAIYFSDEEWEVLTEQQKALYREVMRMNYETVLSLEFPFPKPDMITRLEGEEESQNSDEWQLQGGTSAENEESDVKPPDWPNPMNATSQFPQPQHFDSFGLRLPRDITELPEWSEGYPFYMAMGFPGYDLSADDIAGKFQFSRGMRRSYDAGFKLMVVEYAESTNNCQAAKQFGVLEKNVRDWRKVKPQLQNAHAMRRAFRGPKNGRFALVDQRVAEYVRYMQAKGDPITREAMQLKALEIAQEMNIPEKGFKASLGWCRRMMRRYDLSLRHKVPVPQHLPEDLTEKLVTYQRSVLALRRAHDYEVAQMGNADETPICLEVPSRVTVDNQGEKPVLVKTPGREKLKITAMLGVLADGRKLPPYIILRGTYIPPGKFPSGMEIRCHRYGWMTEDLMQDWLEVVWRRRTGAVPKQRGMLILNGFRGHATDSVKNSMESMNTDMVIIPGGLTSQLQVLDVVVYKPLNDSVRAQYSNWLLAGNLALSPTGNAKKPPLGLFLEWVMVAWNSISSESIVQGFKKCHISSNLEEEDDVLWEIESELPGGGEPPKDCDTESMAESN
RUO
POGK
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali