missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human POMT1 Partial ORF (NP_009102.2, 483 a.a. - 580 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010585-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132856

  • 349.00€ / 10µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT2 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS) and limb-girdle muscular dystrophy type 2K (LGMD2K). Several transcript variants encoding different isoforms have been found for this gene

Sequence: LPDWGYRQLEIVGEKLSRGYHGSTVWNVEEHRYGASQEQRERERELHSPAQVDVSRNLSFMARFSELQWRMLALRSDDSEHKYSSSPLEWVTLDTNIA
Specifica

Specifica

NP_009102.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LPDWGYRQLEIVGEKLSRGYHGSTVWNVEEHRYGASQEQRERERELHSPAQVDVSRNLSFMARFSELQWRMLALRSDDSEHKYSSSPLEWVTLDTNIA
RUO
POMT1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10585
POMT1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ37239/LGMD2K/RT
POMT1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali