missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP1R10 Partial ORF (NP_002705.2, 1 a.a. - 83 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005514-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16133195

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene lies within the major histocompatibility complex class I region on chromosome 6. [provided by RefSeq]

Sequence: MGSGPIDPKELLKGLDSFLNRDGEVKSVDGISKIFSLMKEARKMVSRCTYLNILLQTRSPEILVKFIDVGGYKLLNNWLTYSK
Specifica

Specifica

NP_002705.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.87kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSGPIDPKELLKGLDSFLNRDGEVKSVDGISKIFSLMKEARKMVSRCTYLNILLQTRSPEILVKFIDVGGYKLLNNWLTYSK
RUO
PPP1R10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5514
PPP1R10 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAT53/FB19/PNUTS/PP1R10
PPP1R10
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali