missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP1R3A Partial ORF (NP_002702.1, 3 a.a. - 101 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005506-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16173175

  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The glycogen-associated form of protein phosphatase-1 (PP1) derived from skeletal muscle is a heterodimer composed of a 37-kD catalytic subunit and a 124-kD targeting and regulatory subunit. This gene encodes the regulatory subunit which binds to muscle glycogen with high affinity, thereby enhancing dephosphorylation of glycogen-bound substrates for PP1 such as glycogen synthase and glycogen phosphorylase kinase. [provided by RefSeq]

Sequence: PSEVPSQISKDNFLEVPNLSDSLCEDEEVTFQPGFSPQPSRRGSDSSEDIYLDTPSSGTRRVSFADSFGFNLVSVKEFDCWELPSASTTFDLGTDIFHT
Specifica

Specifica

NP_002702.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PSEVPSQISKDNFLEVPNLSDSLCEDEEVTFQPGFSPQPSRRGSDSSEDIYLDTPSSGTRRVSFADSFGFNLVSVKEFDCWELPSASTTFDLGTDIFHT
RUO
PPP1R3A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5506
PPP1R3A (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GM/PP1G/PPP1R3
PPP1R3A
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali