missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP2R2B Full-length ORF (AAH31790, 1 a.a. - 443 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005521-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129451

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a beta isoform of the regulatory subunit B55 subfamily. Defects in this gene cause autosomal dominant spinocerebellar ataxia 12 (SCA12), a disease caused by degeneration of the cerebellum, sometimes involving the brainstem and spinal cord, and in resulting in poor coordination of speech and body movements. Multiple alternatively spliced variants, which encode different isoforms, have been identified for this gene. The 5' UTR of some of these variants includes a CAG trinucleotide repeat sequence (7-28 copies) that can be expanded to 66-78 copies in cases of SCA12. [provided by RefSeq]

Sequence: MEEDIDTRKINNSFLRDHSYATEADIISTVEFNHTGELLATGDKGGRVVIFQREQESKNQVHRRGEYNVYSTFQSHEPEFDYLKSLEIEEKINKIRWLPQQNAAYFLLSTNDKTVKLWKVSERDKRPEGYNLKDEEGRLRDPATITTLRVPVLRPMDLMVEATPRRVFANAHTYHINSISVNSDYETYMSADDLRINLWNFEITNQSFNIVDIKPANMEELTEVITAAEFHPHHCNTFVYSSSKGTIRLCDMRASALCDRHTKFFEEPEDPSNRSFFSEIISSISDVKFSHSGRYIMTRDYLTVKVWDLNMENRPIETYQVHDYLRSKLCSLYENDCIFDKFECVWNGSDSVIMTGSYNNFFRMFDRNTKRDVTLEASRENSKPRAILKPRKVCVGGKRRKDEISVDSLDFSKKILHTAWHPSENIIAVAATNNLYIFQDKVN
Specifica

Specifica

AAH31790
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.25kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
B55-BETA/FLJ95686/MGC24888/PP2A-B55BETA/PP2A-PR55B/PP2AB-BETA/PP2APR55-BETA/PR2AB-BETA/PR2AB55-BETA/PR2APR55-BETA/PR52B/PR55-BETA/SCA12
PPP2R2B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5521
PPP2R2B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEDIDTRKINNSFLRDHSYATEADIISTVEFNHTGELLATGDKGGRVVIFQREQESKNQVHRRGEYNVYSTFQSHEPEFDYLKSLEIEEKINKIRWLPQQNAAYFLLSTNDKTVKLWKVSERDKRPEGYNLKDEEGRLRDPATITTLRVPVLRPMDLMVEATPRRVFANAHTYHINSISVNSDYETYMSADDLRINLWNFEITNQSFNIVDIKPANMEELTEVITAAEFHPHHCNTFVYSSSKGTIRLCDMRASALCDRHTKFFEEPEDPSNRSFFSEIISSISDVKFSHSGRYIMTRDYLTVKVWDLNMENRPIETYQVHDYLRSKLCSLYENDCIFDKFECVWNGSDSVIMTGSYNNFFRMFDRNTKRDVTLEASRENSKPRAILKPRKVCVGGKRRKDEISVDSLDFSKKILHTAWHPSENIIAVAATNNLYIFQDKVN
RUO
PPP2R2B
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali