missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP3CC Full-length ORF (NP_005596.2, 1 a.a. - 512 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005533-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16171678

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Calmodulin-dependent protein phosphatase, calcineurin, is involved in a wide range of biologic activities, acting as a Ca(2+)-dependent modifier of phosphorylation status. In testis, the motility of the sperm is thought to be controlled by cAMP-dependent phosphorylation and a unique form of calcineurin appears to be associated with the flagellum. The calcineurin holoenzyme is composed of catalytic and regulatory subunits of 60 and 18 kD, respectively. At least 3 genes, calcineurin A-alpha (CALNA1; MIM 114105), calcineurin A-beta (CALNA2; MIM 114106), and calcineurin A-gamma (CALNA3), have been cloned for the catalytic subunit. These genes have been identified in humans, mice, and rats, and are highly conserved between species (90 to 95% amino acid identity).[supplied by OMIM]

Sequence: MSGRRFHLSTTDRVIKAVPFPPTQRLTFKEVFENGKPKVDVLKNHLVKEGRLEEEVALKIINDGAAILRQEKTMIEVDAPITVCGDIHGQFFDLMKLFEVGGSPSNTRYLFLGDYVDRGYFSIECVLYLWSLKINHPKTLFLLRGNHECRHLTDYFTFKQECRIKYSEQVYDACMETFDCLPLAALLNQQFLCVHGGMSPEITSLDDIRKLDRFTEPPAFGPVCDLLWSDPSEDYGNEKTLEHYTHNTVRGCSYFYSYPAVCEFLQNNNLLSIIRAHEAQDAGYRMYRKSQATGFPSLITIFSAPNYLDVYNNKAAVLKYENNVMNIRQFNCSPHPYWLPNFMDVFTWSLPFVGEKVTEMLVNVLNICSDDELISDDEAEGSTTVRKEIIRNKIRAIGKMARVFSILRQESESVLTLKGLTPTGTLPLGVLSGGKQTIETATVEAVEAREAIRGFSLQHKIRSFEEARGLDRINERMPPRKDSIHAGGPMKSVTSAHSHAAHRSDQGKKAHS
Specifica

Specifica

NP_005596.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CALNA3
PPP3CC
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5533
PPP3CC (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSGRRFHLSTTDRVIKAVPFPPTQRLTFKEVFENGKPKVDVLKNHLVKEGRLEEEVALKIINDGAAILRQEKTMIEVDAPITVCGDIHGQFFDLMKLFEVGGSPSNTRYLFLGDYVDRGYFSIECVLYLWSLKINHPKTLFLLRGNHECRHLTDYFTFKQECRIKYSEQVYDACMETFDCLPLAALLNQQFLCVHGGMSPEITSLDDIRKLDRFTEPPAFGPVCDLLWSDPSEDYGNEKTLEHYTHNTVRGCSYFYSYPAVCEFLQNNNLLSIIRAHEAQDAGYRMYRKSQATGFPSLITIFSAPNYLDVYNNKAAVLKYENNVMNIRQFNCSPHPYWLPNFMDVFTWSLPFVGEKVTEMLVNVLNICSDDELISDDEAEGSTTVRKEIIRNKIRAIGKMARVFSILRQESESVLTLKGLTPTGTLPLGVLSGGKQTIETATVEAVEAREAIRGFSLQHKIRSFEEARGLDRINERMPPRKDSIHAGGPMKSVTSAHSHAAHRSDQGKKAHS
RUO
PPP3CC
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali