missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PRKG2 Partial ORF (NP_006250, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005593-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: MGNGSVKPKHSKHPDGHSGNLTTDALRNKVTELERELRRKDAEIQEREYHLKELREQLSKQTVAIAELTEELQNKCIQLNKLQDVVHMQGGSPLQASPDKSpecifica
NP_006250 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGNGSVKPKHSKHPDGHSGNLTTDALRNKVTELERELRRKDAEIQEREYHLKELREQLSKQTVAIAELTEELQNKCIQLNKLQDVVHMQGGSPLQASPDK | |
RUO | |
PRKG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5593 | |
PRKG2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PRKGR2/cGKII | |
PRKG2 | |
Recombinant | |
wheat germ expression system |