missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PSG11 Partial ORF (NP_976032, 126 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005680-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: PRIFPSVTSYYSGENLDLSCFANSNPPAQYSWTINGKFQLSGQKLFIPQITPKHNGLYACSARNSATGEESSTSLTIRVIAPPGLGTFAFNNPTSpecifica
NP_976032 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PRIFPSVTSYYSGENLDLSCFANSNPPAQYSWTINGKFQLSGQKLFIPQITPKHNGLYACSARNSATGEESSTSLTIRVIAPPGLGTFAFNNPT | |
RUO | |
PSG11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5680 | |
PSG11 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC22484/PSG13/PSG14 | |
PSG11 | |
Recombinant | |
wheat germ expression system |