missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PSTPIP1 Full-length ORF (AAH08602, 1 a.a. - 416 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00009051-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16153222

  • 335.00€ / 10µg
Spedizione stimata: 11-06-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene binds to the cytoplasmic tail of CD2, an effector of T cell activation and adhesion, negatively affecting CD2-triggered T cell activation. The encoded protein appears to be a scaffold protein and a regulator of the actin cytoskeleton. It has also been shown to bind ABL1, PTPN18, WAS, CD2AP, and PTPN12. Mutations in this gene are a cause of PAPA syndrome. [provided by RefSeq]

Sequence: MMPQLQFKDAFWCRDFTAHTGYEVLLQRLLDGRKMCKDMEELLRQRAQAEERYGKELVQIARKAGGQTEINSLRASFDSLKQQMENVGSSHIQLALTLREELRSLEEFRERQKEQRKKYEAVMDRVQKSKLSLYKKAMESKKTYEQKCRDADDAEQAFERISANGHQKQVEKSQNKARQCKDSATEAERVYRQSIAQLEKVRAEWEQEHRTTCEAFQLQEFDRLTILRNALWVHSNQLSMQCVKDDELYEEVRLTLEGCSIDADIDSFIQAKSTGTEPPAPVPYQNYYDREVTPLTSSPGIQPSCGMIKRFSGLLHGSPKTTSLAASAASTETLTPTPERNEGVYTAIAVQEIQGNPASPAQEYRALYDYTAQNPDELDLSAGDILEVILEGEDGWWTVERNGQRGFVPGSYLEKL
Specifica

Specifica

AAH08602
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD2BP1/CD2BP1L/CD2BP1S/H-PIP/PAPAS/PSTPIP
PSTPIP1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9051
PSTPIP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMPQLQFKDAFWCRDFTAHTGYEVLLQRLLDGRKMCKDMEELLRQRAQAEERYGKELVQIARKAGGQTEINSLRASFDSLKQQMENVGSSHIQLALTLREELRSLEEFRERQKEQRKKYEAVMDRVQKSKLSLYKKAMESKKTYEQKCRDADDAEQAFERISANGHQKQVEKSQNKARQCKDSATEAERVYRQSIAQLEKVRAEWEQEHRTTCEAFQLQEFDRLTILRNALWVHSNQLSMQCVKDDELYEEVRLTLEGCSIDADIDSFIQAKSTGTEPPAPVPYQNYYDREVTPLTSSPGIQPSCGMIKRFSGLLHGSPKTTSLAASAASTETLTPTPERNEGVYTAIAVQEIQGNPASPAQEYRALYDYTAQNPDELDLSAGDILEVILEGEDGWWTVERNGQRGFVPGSYLEKL
RUO
PSTPIP1
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali