Learn More
ACROBiosystems Human PVRIG Protein, His Tag (MALS verified)
Marca: ACROBiosystems PVG-H52H5-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Human PVRIG (poliovirus receptor related immunoglobulin domain-containing protein), also known as CD112 receptor (CD112R), is an approximately 34 kDa single transmembrane protein in the poliovirus receptor-like protein (PVR) family. The CD112R gene encodes a putative single transmembrane protein, which is composed of a single extracellular IgV domain, one transmembrane domain, and a long intracellular domain. Notably, the intracellular domain of phatases. The extracellular domain sequence of human and mouse CD112R have 65.3% similarity. CD112R may act as a coinhibitory receptor that suppresses T-cell receptor-mediated signals.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.7 kDa. The protein migrates as 22-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
NP_076975.2 | |
1 mg | |
Thr 41 - Asp 171 | |
C7orf15,C7orf15MGC138295,CD112R,MGC104322,MGC138297,MGC2463,PVRIG,CD112 receptor | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |
15.7 kDa | |
Human PVRIG, His Tag (PVG-H52H5) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # Q6DKI7-1). | |
-20°C | |
PVRIG | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |