missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAPGEF1 Partial ORF (AAH41710, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00002889-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16125741

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined. [provided by RefSeq]

Sequence: MGNAIEKQKPLKRSHLYPWKQDSQRSHLSSFTMKLMDKFHSPKIKRTPSKKGKPAEVSVKIPEKPVNKEATDRFLPEGYPLPLDLEQQAVEFMSTSAVASRSQRQKNLSW
Specifica

Specifica

AAH41710
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MGNAIEKQKPLKRSHLYPWKQDSQRSHLSSFTMKLMDKFHSPKIKRTPSKKGKPAEVSVKIPEKPVNKEATDRFLPEGYPLPLDLEQQAVEFMSTSAVASRSQRQKNLSW
RUO
RAPGEF1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2889
RAPGEF1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C3G/DKFZp781P1719/GRF2
RAPGEF1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali