Learn More
Abnova™ Human RASAL1 Full-length ORF (AAH14420.1, 1 a.a. - 776 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008437-P01.10ug
Descrizione
The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. These proteins stimulate the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This particular family member contains domains which are characteristic of the GAP1 subfamily of RasGAP proteins but, in contrast to the other GAP1 family members, this protein is strongly and selectively expressed in endocrine tissues. [provided by RefSeq]
Sequence: MAKSSSLNVRVVEGRALPAKDVSGSSDPYCLVKVDDEVVARTATVWRSLGPFWGEEYTVHLPLDFHQLAFYVLDEDTVGHDDIIGKISLSREAITADPRGIDSWINLSRVDPDAEVQGEICLSVQMLEDGQGRCLRCHVLQARDLAPRDISGTSDPFARVFWGSQSLETSTIKKTRFPHWDEVLELREMPGAPSPLRVELWDWDMVGKNDFLGMVEFSPKTLQQKPPKGWFRLLPFPRAEEDSGGNLGALRVKVRLIEDRVLPSQCYQPLMELLMESVQGPAEEDTASPLALLEELTLGDCRQDLATKLVKLFLGRGLAGHFLDYLTRREVARTMDPNTLFRSNSLASKSMEQFMKLVGMPYLHEVLKPVISRVFEEKKYMELDPCKMDLGRTRRISFKGALSEEQMRETSLGLLTGYLGPIVDAIVGSVGRCPPAMRLAFKQLHRRVEERFPQAEHQDVKYLAISGFLFLRFFAPAILTPKLFDLRDQHADPQTSRSLLLLAKAVQSIGNLGQQLGQGKELWMAPLHPFLLQCVSRVRDFLDRLVDVDGDEAGVPARALFPPSAIVREGYLLKRKEEPAGLATRFAFKKRYVWLSGETLSFSKSPEWQVVTQDGTGALHTTYLQCKNVNELNQWLSALRKASAPNPNKLAACHPGAFRSARWTCCLQAERSAAGCSRTHSAVTLGDWSDPLDPDAEAQTVYRQLLLGRDQLRLKLLEDSNMDTTLEADTGACPEVLARQRAATARLLEVLADLDRAHEEFQQQERGKAALGPLGPSpecifica
AAH14420.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
113.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RASAL | |
RASAL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8437 | |
RASAL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKSSSLNVRVVEGRALPAKDVSGSSDPYCLVKVDDEVVARTATVWRSLGPFWGEEYTVHLPLDFHQLAFYVLDEDTVGHDDIIGKISLSREAITADPRGIDSWINLSRVDPDAEVQGEICLSVQMLEDGQGRCLRCHVLQARDLAPRDISGTSDPFARVFWGSQSLETSTIKKTRFPHWDEVLELREMPGAPSPLRVELWDWDMVGKNDFLGMVEFSPKTLQQKPPKGWFRLLPFPRAEEDSGGNLGALRVKVRLIEDRVLPSQCYQPLMELLMESVQGPAEEDTASPLALLEELTLGDCRQDLATKLVKLFLGRGLAGHFLDYLTRREVARTMDPNTLFRSNSLASKSMEQFMKLVGMPYLHEVLKPVISRVFEEKKYMELDPCKMDLGRTRRISFKGALSEEQMRETSLGLLTGYLGPIVDAIVGSVGRCPPAMRLAFKQLHRRVEERFPQAEHQDVKYLAISGFLFLRFFAPAILTPKLFDLRDQHADPQTSRSLLLLAKAVQSIGNLGQQLGQGKELWMAPLHPFLLQCVSRVRDFLDRLVDVDGDEAGVPARALFPPSAIVREGYLLKRKEEPAGLATRFAFKKRYVWLSGETLSFSKSPEWQVVTQDGTGALHTTYLQCKNVNELNQWLSALRKASAPNPNKLAACHPGAFRSARWTCCLQAERSAAGCSRTHSAVTLGDWSDPLDPDAEAQTVYRQLLLGRDQLRLKLLEDSNMDTTLEADTGACPEVLARQRAATARLLEVLADLDRAHEEFQQQERGKAALGPLGP | |
RUO | |
RASAL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |