missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RASEF Partial ORF (AAH23566, 631 a.a. - 740 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00158158-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: NIREWVDMIEDAAHETVPIMLVGNKADIRDTAATEGQKCVPGHFGEKLAMTYGALFCETSAKDGSNIVEAVLHLAREVKKRTDKDDSRSITNLTGTNSKKSPQMKNCCNGSpecifica
AAH23566 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NIREWVDMIEDAAHETVPIMLVGNKADIRDTAATEGQKCVPGHFGEKLAMTYGALFCETSAKDGSNIVEAVLHLAREVKKRTDKDDSRSITNLTGTNSKKSPQMKNCCNG | |
RUO | |
RASEF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
158158 | |
RASEF (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31614/RAB45 | |
RASEF | |
Recombinant | |
wheat germ expression system |