missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RBBP6 Full-length ORF (NP_116015.2, 1 a.a. - 118 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005930-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172168

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The retinoblastoma tumor suppressor (pRB) protein binds with many other proteins. In various human cancers, pRB suppresses cellular proliferation and is inactivated. Cell cycle-dependent phosphorylation regulates the activity of pRB. This gene encodes a protein which binds to underphosphorylated but not phosphorylated pRB. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MSCVHYKFSSKLNYDTVTFDGLHISLCDLKKQIMGREKLKAADCDLQITNAQTKEEYTDDNALIPKNSSVIVRRIPIGGVKSTSKTYVISRTEPAMATTKAVCKNTISHFFYTLLLPL
Specifica

Specifica

NP_116015.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686P0638/DKFZp761B2423/MY038/P2P-R/PACT/RBQ-1/SNAMA
RBBP6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5930
RBBP6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSCVHYKFSSKLNYDTVTFDGLHISLCDLKKQIMGREKLKAADCDLQITNAQTKEEYTDDNALIPKNSSVIVRRIPIGGVKSTSKTYVISRTEPAMATTKAVCKNTISHFFYTLLLPL
RUO
RBBP6
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali