missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RBBP7 Full-length ORF (NP_002884.1, 1 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005931-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16130112

  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation. [provided by RefSeq]

Sequence: MASKEMFEDTVEERVINEEYKIWKKNTPFLYDLVMTHALQWPSLTVQWLPEVTKPEGKDYALHWLVLGTHTSDEQNHLVVARVHIPNDDAQFDASHCDSDKGEFGGFGSVTGKIECEIKINHEGEVNRARYMPQNPHIIATKTPSSDVLVFDYTKHPAKPDPSGECNPDLRLRGHQKEGYGLSWNSNLSGHLLSASDDHTVCLWDINAGPKEGKIVDAKAIFTGHSAVVEDVAWHLLHESLFGSVADDQKLMIWDTRSNTTSKPSHLVDAHTAEVNCLSFNPYSEFILATGSADKTVALWDLRNLKLKLHTFESHKDEIFQVHWSPHNETILASSGTDRRLNVWDLSKIGEEQSAEDAEDGPPELLFIHGGHTAKISDFSWNPNEPWVICSVSEDNIMQIWQMAENIYNDEESDVTTSELEGQGS
Specifica

Specifica

NP_002884.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC138867/MGC138868/RbAp46
RBBP7
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5931
RBBP7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASKEMFEDTVEERVINEEYKIWKKNTPFLYDLVMTHALQWPSLTVQWLPEVTKPEGKDYALHWLVLGTHTSDEQNHLVVARVHIPNDDAQFDASHCDSDKGEFGGFGSVTGKIECEIKINHEGEVNRARYMPQNPHIIATKTPSSDVLVFDYTKHPAKPDPSGECNPDLRLRGHQKEGYGLSWNSNLSGHLLSASDDHTVCLWDINAGPKEGKIVDAKAIFTGHSAVVEDVAWHLLHESLFGSVADDQKLMIWDTRSNTTSKPSHLVDAHTAEVNCLSFNPYSEFILATGSADKTVALWDLRNLKLKLHTFESHKDEIFQVHWSPHNETILASSGTDRRLNVWDLSKIGEEQSAEDAEDGPPELLFIHGGHTAKISDFSWNPNEPWVICSVSEDNIMQIWQMAENIYNDEESDVTTSELEGQGS
RUO
RBBP7
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali