Learn More
Abnova™ Human RGS11 Full-length ORF (AAI53020.1, 1 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008786-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene belongs to the RGS (regulator of G protein signaling) family. Members of the RGS family act as GTPase-activating proteins on the alpha subunits of heterotrimeric, signal-transducing G proteins. This protein inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits, thereby driving them into their inactive GDP-bound form. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]
Sequence: MAAGPAPPPGRPRAQMPHLRKMERVVVSMQDPDQGVKMRSQRLLVTVIPHAVTGSDVVQWLAQKFCVSEEEALHLGAVLVQHGYIYPLRDPRSLMLRPDETPYRFQTPYFWTSTLRPAAELDYAIYLAKKNIRKRGTLVDYEKDCYDRLHKKINHAWDLVLMQAREQLRAAKQRSKGDRLVIACQEQTYWLVNRPPPGAPDVLEQGPGRGSCAASRVLMTKSADFHKREIEYFRKALGRTRVKSSVCLEAYLSFCGQRGPHDPLVSGCLPSNPWISDNDAYWVMNAPTVAAPTKLRVERWGFSFRELLEDPVGRAHFMDFLGKEFSGENLSFWEACEELRYGAQAQVPTLVDAVYEQFLAPGAAHWVNIDSRTMEQTLEGLRQPHRYVLDDAQLHIYMLMKKDSYPRFLKSDMYKALLAEAGIPLEMKRRVFPFTWRPRHSSPSPALLPTPVEPTAACGPGGGDGVASpecifica
AAI53020.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.32kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RS11 | |
RGS11 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8786 | |
RGS11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAGPAPPPGRPRAQMPHLRKMERVVVSMQDPDQGVKMRSQRLLVTVIPHAVTGSDVVQWLAQKFCVSEEEALHLGAVLVQHGYIYPLRDPRSLMLRPDETPYRFQTPYFWTSTLRPAAELDYAIYLAKKNIRKRGTLVDYEKDCYDRLHKKINHAWDLVLMQAREQLRAAKQRSKGDRLVIACQEQTYWLVNRPPPGAPDVLEQGPGRGSCAASRVLMTKSADFHKREIEYFRKALGRTRVKSSVCLEAYLSFCGQRGPHDPLVSGCLPSNPWISDNDAYWVMNAPTVAAPTKLRVERWGFSFRELLEDPVGRAHFMDFLGKEFSGENLSFWEACEELRYGAQAQVPTLVDAVYEQFLAPGAAHWVNIDSRTMEQTLEGLRQPHRYVLDDAQLHIYMLMKKDSYPRFLKSDMYKALLAEAGIPLEMKRRVFPFTWRPRHSSPSPALLPTPVEPTAACGPGGGDGVA | |
RUO | |
RGS11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |