Learn More
Abnova™ Human RHOBTB1 Partial ORF (AAH32848, 494 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009886-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene belongs to the Rho family of the small GTPase superfamily. It contains a GTPase domain, a proline-rich region, a tandem of 2 BTB (broad complex, tramtrack, and bric-a-brac) domains, and a conserved C-terminal region. The protein plays a role in small GTPase-mediated signal transduction and the organization of the actin filament system. Alternate splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: GAISAHKPLLICSCEWMAAMFGGSFVESANSEVYLPNINKISMQAVLDYLYTKQLSPNLDLDPLELIALANRFCLPHLVALAEQHAVQELTKAATSGVGIDGEVLSYLELSpecifica
AAH32848 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GAISAHKPLLICSCEWMAAMFGGSFVESANSEVYLPNINKISMQAVLDYLYTKQLSPNLDLDPLELIALANRFCLPHLVALAEQHAVQELTKAATSGVGIDGEVLSYLEL | |
RUO | |
RHOBTB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9886 | |
RHOBTB1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0740/MGC33059/MGC33841 | |
RHOBTB1 | |
Recombinant | |
wheat germ expression system |