Learn More
Abnova™ Human ROPN1B Full-length ORF (NP_001012337.1, 1 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00152015-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16128713
  • 508.00€ / 25µg
Spedizione stimata: 28-08-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Sequence: MAQTDKPTCIPPELPKMLKEFAKAAIRAQPQDLIQWGADYFEALSRGETPPVRERSERVALCNWAELTPELLKILHSQVAGRLIIRAEELAQMWKVVNLPTDLFNSVMNVGRFTEEIEWLKFLALACSALGVTITKTLKIVCEVLSCDHNGGLPRIPFSTFQFLYTYIAEVDGEICASHVSRMLNYIEQEVIGPDGLITVNDFTQNPRVWLE
Specifica
Specifica
NP_001012337.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ROPN1B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
152015
ROPN1B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAQTDKPTCIPPELPKMLKEFAKAAIRAQPQDLIQWGADYFEALSRGETPPVRERSERVALCNWAELTPELLKILHSQVAGRLIIRAEELAQMWKVVNLPTDLFNSVMNVGRFTEEIEWLKFLALACSALGVTITKTLKIVCEVLSCDHNGGLPRIPFSTFQFLYTYIAEVDGEICASHVSRMLNYIEQEVIGPDGLITVNDFTQNPRVWLE
RUO
ROPN1B
Recombinant
wheat germ expression system