missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RRP15 Full-length ORF (AAH20641.1, 1 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00051018-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129512

  • 349.00€ / 10µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a protein that co-purifies with human nucleoli. A similar protein in budding yeast is a component of pre-60S ribosomal particles, and is required for the early maturation steps of the 60S subunit. [provided by RefSeq]

Sequence: MKMVTGAVASVLEDEATDTSDSEGSCGSEKDHFYSDDDAIEADSEGDAEPCDKENENDGESSVGTNMGWADAMAKVLNKKTPESKPTILVKNKKLEKEKEKLKQERLEKIKQRDKRLEWEMMCRVKPDVVQDKETERNLQRIATRGVVQLFNAVQKHQKNVDEKVKEAGSSMRKRAKLISTVSKKDFISVLRGMDGSTNETASSRKKPKAKQTEVKSEEGPGWTILRDDFMMGASMKDWDKESDGPDDSRPESASDSDT
Specifica

Specifica

AAH20641.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CGI-115/KIAA0507/MGC22291
RRP15
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51018
RRP15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKMVTGAVASVLEDEATDTSDSEGSCGSEKDHFYSDDDAIEADSEGDAEPCDKENENDGESSVGTNMGWADAMAKVLNKKTPESKPTILVKNKKLEKEKEKLKQERLEKIKQRDKRLEWEMMCRVKPDVVQDKETERNLQRIATRGVVQLFNAVQKHQKNVDEKVKEAGSSMRKRAKLISTVSKKDFISVLRGMDGSTNETASSRKKPKAKQTEVKSEEGPGWTILRDDFMMGASMKDWDKESDGPDDSRPESASDSDT
RUO
RRP15
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali