missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human S100B Full-length ORF (NP_006263.1, 1 a.a. - 92 a.a.) Recombinant Protein MW: 37.1kDa with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00006285-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132518

  • 528.00€ / 25µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21; however, this gene is located at 21q22.3. This protein may function in Neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several neurological, neoplastic, and other types of diseases, including Alzheimer's disease, Down's syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes. [provided by RefSeq]

Sequence: MSELEKAMVALIDVFHQYSGREGDKHKLKKSELKELINNELSHFLEEIKEQEVVDKVMETLDNDGDGECDFQEFMAFVAMVTTACHEFFEHE
Specifica

Specifica

NP_006263.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NEF/S100/S100beta
S100B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6285
S100B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSELEKAMVALIDVFHQYSGREGDKHKLKKSELKELINNELSHFLEEIKEQEVVDKVMETLDNDGDGECDFQEFMAFVAMVTTACHEFFEHE
RUO
S100B
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali